Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYO18B
Entrez Gene ID
84700
Gene Name
myosin XVIIIB
Gene Aliases
KFS4
Chromosome Location
Chr.22: 25742133 - 26063847 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318245.1 NP_001305174.1 5 - 6 1832 60
NM_032608.6 NP_115997.5 5 - 6 1832 60
XM_011530458.2 XP_011528760.1 6 - 7 1968 60
XM_011530459.2 XP_011528761.1 6 - 7 1939 60
XM_011530460.2 XP_011528762.1 5 - 6 1812 60
XM_011530461.2 XP_011528763.1 6 - 7 1968 60
XM_011530464.2 XP_011528766.1 6 - 7 1968 60
XM_011530465.2 XP_011528767.1 5 - 6 1725 60
XM_011530466.2 XP_011528768.1 6 - 7 1969 60
XM_017029012.1 XP_016884501.1 6 - 7 1968 60
XM_017029013.1 XP_016884502.1 6 - 7 2016 60
XM_017029014.1 XP_016884503.1 6 - 7 1968 60
XM_017029015.1 XP_016884504.1 5 - 6 1842 60
XM_017029017.1 XP_016884506.1 5 - 6 1841 60
GenBank mRNA AB042648.2 - 1 - 2 144 60
AB075376.1 - 5 - 6 1832 60
AJ310931.2 - 5 - 6 1823 60
AY077700.1 - 5 - 6 1742 60
BC144596.1 - 4 - 5 1632 60
BC144597.1 - 4 - 5 1632 60
BC150626.1 - 4 - 5 1632 60

View Full Product Details