The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_011537317.2 | XP_011535619.1 | 10 - 11 | 1174 | 157 |
XM_011537318.2 | XP_011535620.1 | 10 - 11 | 1174 | 157 | |
XM_017021760.1 | XP_016877249.1 | 9 - 10 | 877 | 157 | |
XM_017021761.1 | XP_016877250.1 | 4 - 5 | 330 | 157 | |
GenBank mRNA | AK000913.1 | - | 10 - 11 | 905 | 157 |
AK293236.1 | - | 9 - 10 | 789 | 157 | |
AK295010.1 | - | 9 - 10 | 839 | 157 | |
AK304695.1 | - | 10 - 11 | 910 | 157 | |
AK308537.1 | - | 8 - 9 | 687 | 157 | |
BC006001.1 | - | 9 - 10 | 791 | 157 | |
BC056687.1 | - | 10 - 11 | 921 | 157 |