Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- SCLY
- Entrez Gene ID
-
51540
- Gene Name
- selenocysteine lyase
- Gene Aliases
-
SCL, hSCL
- Chromosome Location
-
Chr.2: 238060924 - 238099413 on Build GRCh38
- Gene Symbol
- UBE2F-SCLY
- Entrez Gene ID
-
100533179
- Gene Name
- UBE2F-SCLY readthrough (NMD candidate)
- Gene Aliases
-
-
- Chromosome Location
-
Chr.2: 237966945 - 238099413 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_037904.1 |
|
10 - 11 |
878 |
106 |
View Full Product Details