Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRPC7
Entrez Gene ID
57113
Gene Name
transient receptor potential cation channel subfamily C member 7
Gene Aliases
TRP7
Chromosome Location
Chr.5: 136213311 - 136365476 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001167576.1 NP_001161048.1 5 - 6 1719 80
NM_001167577.1 NP_001161049.1 6 - 7 1884 80
NM_020389.2 NP_065122.1 7 - 8 2067 80
GenBank mRNA AJ272034.1 - 6 - 7 1845 80
AJ421783.1 - 6 - 7 1845 80
AJ549088.1 - 5 - 6 1662 80
AJ549089.1 - 5 - 6 1680 80
AJ549090.1 - 3 - 4 1280 80
AY167926.1 - 5 - 6 1677 80
AY167927.1 - 4 - 5 1497 80
BC128185.1 - 6 - 7 1845 80
BC128186.1 - 6 - 7 1861 80

View Full Product Details