Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BZW2
Entrez Gene ID
28969
Gene Name
basic leucine zipper and W2 domains 2
Gene Aliases
HSPC028, MST017, MSTP017
Chromosome Location
Chr.7: 16646134 - 16706523 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159767.1 NP_001153239.1 5 - 6 582 118
NM_014038.2 NP_054757.1 5 - 6 566 118
XM_006715706.1 XP_006715769.1 5 - 6 450 118
XM_006715707.1 XP_006715770.1 5 - 6 533 118
XM_006715708.1 XP_006715771.1 5 - 6 452 118
GenBank mRNA AF083246.1 - 5 - 6 566 118
AF110323.1 - 4 - 5 404 118
AK001218.1 - 5 - 6 546 118
AK001260.1 - 5 - 6 540 118
AK027837.1 - 5 - 6 450 118
AK297496.1 - 5 - 6 495 118
BC003056.1 - 5 - 6 492 118
BC008453.1 - 5 - 6 519 118
BC009597.1 - 5 - 6 550 118
BC107757.1 - 5 - 6 540 118

View Full Product Details