The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000142.4 | NP_000133.1 | 9 - 10 | 1524 | 150 |
NM_001163213.1 | NP_001156685.1 | 9 - 10 | 1530 | 150 | |
XM_006713871.1 | XP_006713934.1 | 9 - 10 | 1543 | 150 | |
XM_006713873.1 | XP_006713936.1 | 9 - 10 | 1537 | 150 | |
GenBank mRNA | BC121175.2 | - | 2 - 3 | 291 | 150 |
BC128610.1 | - | 2 - 3 | 291 | 150 | |
BC153824.1 | - | 2 - 3 | 291 | 150 | |
HM244683.1 | - | 9 - 10 | 1419 | 150 | |
M58051.1 | - | 8 - 9 | 1307 | 150 | |
M64347.1 | - | 7 - 8 | 1043 | 150 |