Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNRNP40
Entrez Gene ID
9410
Gene Name
small nuclear ribonucleoprotein U5 subunit 40
Gene Aliases
40K, HPRP8BP, PRP8BP, PRPF8BP, SPF38, WDR57
Chromosome Location
Chr.1: 31259568 - 31296797 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004814.2 NP_004805.2 2 - 3 317 74
GenBank mRNA AF090988.1 - 2 - 3 286 74
AK074516.1 - 2 - 3 300 74
AK298823.1 - 2 - 3 335 74
BC001494.2 - 2 - 3 293 74
CR736901.1 - 2 - 3 317 74
EF011621.1 - 2 - 3 292 74

View Full Product Details