Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SNRNP40
Entrez Gene ID
9410
Gene Name
small nuclear ribonucleoprotein U5 subunit 40
Gene Aliases
40K, HPRP8BP, PRP8BP, PRPF8BP, SPF38, WDR57
Chromosome Location
Chr.1: 31259568 - 31296797 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004814.2 NP_004805.2 4 - 5 573 132
GenBank mRNA AF090988.1 - 4 - 5 542 132
AK074516.1 - 4 - 5 556 132
AK298823.1 - 4 - 5 591 132
AK313800.1 - 4 - 5 538 132
BC001494.2 - 4 - 5 549 132
EF011621.1 - 4 - 5 548 132

View Full Product Details