The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030816.4 | NP_110443.3 | 5 - 6 | 1037 | 86 |
XM_005271234.3 | XP_005271291.1 | 5 - 6 | 1280 | 86 | |
XM_005271235.3 | XP_005271292.1 | 5 - 6 | 1027 | 86 | |
XM_006710929.3 | XP_006710992.1 | 5 - 6 | 1050 | 86 | |
XM_017002413.1 | XP_016857902.1 | 3 - 4 | 260 | 86 | |
GenBank mRNA | AL136717.1 | - | 4 - 5 | 933 | 86 |
BC028840.1 | - | 5 - 6 | 1026 | 86 |