Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLIN3
Entrez Gene ID
10226
Gene Name
perilipin 3
Gene Aliases
M6PRBP1, PP17, TIP47
Chromosome Location
Chr.19: 4838334 - 4867768 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164189.1 NP_001157661.1 5 - 6 814 119
NM_001164194.1 NP_001157666.1 5 - 6 778 119
NM_005817.4 NP_005808.3 5 - 6 814 119
GenBank mRNA AF051314.1 - 1 - 2 126 119
AF051315.1 - 1 - 2 102 119
AF055574.1 - 5 - 6 690 119
AF057140.1 - 5 - 6 711 119
AK122671.1 - 5 - 6 665 119
AK223054.1 - 5 - 6 744 119
AK225045.1 - 5 - 6 680 119
AK225400.1 - 5 - 6 695 119
AK226088.1 - 5 - 6 692 119
AK291104.1 - 5 - 6 771 119
AK312740.1 - 5 - 6 740 119
BC001590.2 - 5 - 6 687 119
BC005818.2 - 5 - 6 703 119
BC007566.1 - 5 - 6 703 119
BC019278.1 - 5 - 6 703 119
BQ063091.1 - 4 - 5 668 119
BT007235.1 - 4 - 5 637 119

View Full Product Details