Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MMEL1
Entrez Gene ID
79258
Gene Name
membrane metallo-endopeptidase-like 1
Gene Aliases
MMEL2, NEP2, NEPII, NL1, NL2, SEP
Chromosome Location
Chr.1: 2590639 - 2633042 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033467.3 NP_258428.2 3 - 4 450 88
XM_017002310.1 XP_016857799.1 4 - 5 716 88
XM_017002311.1 XP_016857800.1 3 - 4 450 88
XM_017002313.1 XP_016857802.1 3 - 4 450 88
XM_017002314.1 XP_016857803.1 3 - 4 450 88
XM_017002315.1 XP_016857804.1 3 - 4 326 88
GenBank mRNA AF336981.1 - 2 - 3 273 88
BC101027.2 - 2 - 3 246 88
BC101028.1 - 2 - 3 246 88
BC101029.2 - 2 - 3 246 88
BC101030.1 - 2 - 3 246 88
DB089501.1 - 3 - 4 450 88

View Full Product Details