Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANO1
Entrez Gene ID
55107
Gene Name
anoctamin 1
Gene Aliases
DOG1, ORAOV2, TAOS2, TMEM16A
Chromosome Location
Chr.11: 70078302 - 70189545 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018043.5 NP_060513.5 24 - 25 2891 88
NR_030691.1 22 - 23 2649 88
XM_006718602.2 XP_006718665.1 25 - 26 2673 88
XM_006718604.2 XP_006718667.1 25 - 26 2788 88
XM_006718605.2 XP_006718668.1 22 - 23 2644 88
XM_011545121.2 XP_011543423.1 25 - 26 2665 88
XM_011545123.2 XP_011543425.1 25 - 26 2662 88
XM_011545124.2 XP_011543426.1 24 - 25 2653 88
XM_011545125.2 XP_011543427.1 24 - 25 2587 88
XM_011545126.2 XP_011543428.1 23 - 24 2575 88
XM_011545127.2 XP_011543429.1 23 - 24 2521 88
XM_011545128.2 XP_011543430.1 22 - 23 2509 88
XM_011545129.2 XP_011543431.1 25 - 26 2580 88
XM_017017956.1 XP_016873445.1 24 - 25 2596 88
XM_017017957.1 XP_016873446.1 23 - 24 2587 88
GenBank mRNA AK001123.1 - 14 - 15 1429 88
AK293088.1 - 24 - 25 2599 88
AK303971.1 - 4 - 5 593 88
AL833582.1 - 24 - 25 2635 88
AY728143.1 - 24 - 25 2891 88
BC027590.2 - 18 - 19 1765 88
BC033036.2 - 22 - 23 2605 88

View Full Product Details