Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANO1
Entrez Gene ID
55107
Gene Name
anoctamin 1
Gene Aliases
DOG1, ORAOV2, TAOS2, TMEM16A
Chromosome Location
Chr.11: 70078302 - 70189545 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018043.5 NP_060513.5 19 - 20 2261 78
NR_030691.1 17 - 18 2019 78
XM_006718602.2 XP_006718665.1 20 - 21 2043 78
XM_006718604.2 XP_006718667.1 20 - 21 2158 78
XM_006718605.2 XP_006718668.1 17 - 18 2014 78
XM_011545121.2 XP_011543423.1 20 - 21 2035 78
XM_011545123.2 XP_011543425.1 20 - 21 2032 78
XM_011545124.2 XP_011543426.1 19 - 20 2023 78
XM_011545125.2 XP_011543427.1 19 - 20 1957 78
XM_011545126.2 XP_011543428.1 18 - 19 1945 78
XM_011545127.2 XP_011543429.1 18 - 19 1891 78
XM_011545128.2 XP_011543430.1 17 - 18 1879 78
XM_011545129.2 XP_011543431.1 20 - 21 1950 78
XM_017017956.1 XP_016873445.1 19 - 20 1966 78
XM_017017957.1 XP_016873446.1 18 - 19 1957 78
GenBank mRNA AB845669.1 - 19 - 20 1956 78
AK001123.1 - 9 - 10 799 78
AK293088.1 - 19 - 20 1969 78
AL833582.1 - 19 - 20 2005 78
AY728143.1 - 19 - 20 2261 78
BC027590.2 - 13 - 14 1135 78
BC033036.2 - 17 - 18 1975 78

View Full Product Details