The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003337.3 | NP_003328.1 | 1 - 2 | 461 | 72 |
GenBank mRNA | AK312012.1 | - | 1 - 2 | 125 | 72 |
BC005979.1 | - | 1 - 2 | 68 | 72 | |
BC008404.1 | - | 1 - 2 | 128 | 72 | |
BC008470.1 | - | 1 - 2 | 128 | 72 | |
BT007071.1 | - | 1 - 2 | 41 | 72 | |
CR407634.1 | - | 1 - 2 | 41 | 72 | |
DA523817.1 | - | 1 - 2 | 460 | 72 | |
M74525.1 | - | 1 - 2 | 462 | 72 | |
X53251.1 | - | 1 - 2 | 79 | 72 |