The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019604.3 | NP_062550.2 | 5 - 6 | 704 | 93 |
XM_011542900.2 | XP_011541202.1 | 4 - 5 | 630 | 93 | |
GenBank mRNA | AF001622.1 | - | 5 - 6 | 655 | 93 |
AK313578.1 | - | 5 - 6 | 702 | 93 | |
BC070266.1 | - | 5 - 6 | 704 | 93 |