The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001010939.2 | NP_001010939.2 | 1 - 2 | 85 | 118 |
GenBank mRNA | BC031219.1 | - | 1 - 2 | 85 | 118 |