The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001010939.2 | NP_001010939.2 | 4 - 5 | 447 | 148 |
XM_006717635.2 | XP_006717698.1 | 4 - 5 | 445 | 148 | |
XM_011539314.2 | XP_011537616.1 | 4 - 5 | 970 | 148 | |
XM_011539315.2 | XP_011537617.1 | 4 - 5 | 446 | 148 | |
XM_011539317.2 | XP_011537619.1 | 4 - 5 | 970 | 148 | |
XM_017015740.1 | XP_016871229.1 | 4 - 5 | 970 | 148 | |
GenBank mRNA | BC031219.1 | - | 4 - 5 | 447 | 148 |