The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152492.2 | NP_689705.2 | 1 - 2 | 400 | 55 |
XM_011540780.1 | XP_011539082.1 | 1 - 2 | 403 | 55 | |
XM_017000402.1 | XP_016855891.1 | 1 - 2 | 403 | 55 | |
XM_017000403.1 | XP_016855892.1 | 1 - 2 | 403 | 55 | |
XM_017000404.1 | XP_016855893.1 | 1 - 2 | 403 | 55 | |
XM_017000405.1 | XP_016855894.1 | 1 - 2 | 403 | 55 | |
GenBank mRNA | AK057387.1 | - | 1 - 2 | 403 | 55 |
BC112108.1 | - | 1 - 2 | 401 | 55 | |
BC112112.1 | - | 1 - 2 | 400 | 55 | |
DB048639.1 | - | 1 - 2 | 358 | 55 |