The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144701.2 | NP_653302.2 | 9 - 10 | 1237 | 162 |
XM_005270516.2 | XP_005270573.1 | 6 - 7 | 883 | 162 | |
XM_011540790.2 | XP_011539092.1 | 9 - 10 | 1705 | 162 | |
XM_011540791.2 | XP_011539093.1 | 9 - 10 | 1222 | 162 | |
GenBank mRNA | AF461422.1 | - | 9 - 10 | 1237 | 162 |
AM990313.1 | - | 6 - 7 | 923 | 162 | |
AM990314.1 | - | 5 - 6 | 553 | 162 | |
AM990315.1 | - | 5 - 6 | 568 | 162 | |
AM990316.1 | - | 5 - 6 | 557 | 162 | |
AM990317.1 | - | 5 - 6 | 624 | 162 | |
AM990319.1 | - | 4 - 5 | 407 | 162 | |
AM990320.1 | - | 4 - 5 | 411 | 162 | |
AM990322.1 | - | 3 - 4 | 250 | 162 | |
AM990323.1 | - | 6 - 7 | 709 | 162 | |
AM990324.1 | - | 6 - 7 | 643 | 162 | |
AM990326.1 | - | 5 - 6 | 331 | 162 | |
AM990327.1 | - | 4 - 5 | 463 | 162 | |
AM990329.1 | - | 3 - 4 | 317 | 162 | |
AM990333.1 | - | 4 - 5 | 406 | 162 | |
AM990336.1 | - | 5 - 6 | 619 | 162 | |
AY937250.1 | - | 7 - 8 | 1044 | 162 | |
AY937251.1 | - | 6 - 7 | 883 | 162 | |
AY937252.1 | - | 6 - 7 | 883 | 162 | |
BC016829.1 | - | 4 - 5 | 440 | 162 |