The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000212.2 | NP_000203.2 | 7 - 8 | 1059 | 126 |
GenBank mRNA | AY826979.1 | - | 7 - 8 | 1048 | 126 |
BC127666.1 | - | 7 - 8 | 1059 | 126 | |
BC127667.1 | - | 7 - 8 | 1063 | 126 | |
J02703.1 | - | 7 - 8 | 1059 | 126 | |
KC120777.1 | - | 7 - 8 | 1059 | 126 | |
KC120778.1 | - | 7 - 8 | 1059 | 126 | |
M20311.1 | - | 7 - 8 | 1055 | 126 | |
M35999.1 | - | 7 - 8 | 1057 | 126 | |
U95204.1 | - | 7 - 8 | 1039 | 126 |