Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CPNE6
Entrez Gene ID
9362
Gene Name
copine 6
Gene Aliases
-
Chromosome Location
Chr.14: 24070837 - 24078100 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001280558.1 NP_001267487.1 17 - 18 91
NM_006032.3 NP_006023.1 16 - 17 91
XM_005268216.1 XP_005268273.1 17 - 18 91
XM_005268217.1 XP_005268274.1 15 - 16 91
GenBank mRNA AB009288.1 - 16 - 17 91
AK125195.1 - 15 - 16 91
AK293655.1 - 17 - 18 91
AL137688.1 - 8 - 9 91
AW157471.1 - 1 - 2 212 91
BC018046.1 - 16 - 17 91
BX118572.1 - 1 - 2 229 91

View Full Product Details