The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190267.1 | NP_001177196.1 | 1 - 2 | 377 | 71 |
NM_017974.3 | NP_060444.3 | 1 - 2 | 377 | 71 | |
NM_030803.6 | NP_110430.5 | 1 - 2 | 377 | 71 | |
NM_198890.2 | NP_942593.2 | 1 - 2 | 377 | 71 | |
XM_005246082.2 | XP_005246139.1 | 1 - 2 | 314 | 71 | |
XM_005246084.2 | XP_005246141.1 | 1 - 2 | 285 | 71 | |
XM_005246086.2 | XP_005246143.1 | 1 - 2 | 285 | 71 | |
XM_006712608.2 | XP_006712671.1 | 1 - 2 | 377 | 71 | |
GenBank mRNA | AK000897.1 | - | 1 - 2 | 276 | 71 |
AK001690.1 | - | 1 - 2 | 223 | 71 | |
AK024453.1 | - | 1 - 2 | 247 | 71 | |
AK123876.1 | - | 1 - 2 | 276 | 71 | |
AK308400.1 | - | 1 - 2 | 277 | 71 | |
AY358182.1 | - | 1 - 2 | 266 | 71 | |
AY398617.1 | - | 1 - 2 | 266 | 71 | |
BC117337.1 | - | 1 - 2 | 39 | 71 | |
BC143900.1 | - | 1 - 2 | 39 | 71 | |
BG702756.1 | - | 1 - 2 | 299 | 71 | |
DB084962.1 | - | 1 - 2 | 377 | 71 | |
DB474638.1 | - | 1 - 2 | 331 | 71 | |
EF079889.1 | - | 1 - 2 | 120 | 71 | |
EF079890.1 | - | 1 - 2 | 120 | 71 | |
EF560734.1 | - | 1 - 2 | 247 | 71 | |
JQ924062.1 | - | 1 - 2 | 120 | 71 |