The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017974.3 | NP_060444.3 | 7 - 8 | 1056 | 60 |
NM_198890.2 | NP_942593.2 | 4 - 5 | 624 | 60 | |
GenBank mRNA | AK001690.1 | - | 7 - 8 | 902 | 60 |
AK027854.1 | - | 6 - 7 | 638 | 60 | |
AK308400.1 | - | 7 - 8 | 956 | 60 | |
BC071846.1 | - | 4 - 5 | 462 | 60 | |
BC143900.1 | - | 7 - 8 | 718 | 60 | |
EF079889.1 | - | 7 - 8 | 799 | 60 | |
EF079890.1 | - | 4 - 5 | 367 | 60 | |
EF560734.1 | - | 7 - 8 | 926 | 60 | |
JQ924062.1 | - | 7 - 8 | 799 | 60 |