The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_006713666.2 | XP_006713729.1 | 1 - 2 | 430 | 69 |
XM_006713668.3 | XP_006713731.1 | 1 - 2 | 439 | 69 | |
XM_006713670.2 | XP_006713733.1 | 1 - 2 | 430 | 69 | |
XM_017006636.1 | XP_016862125.1 | 1 - 2 | 430 | 69 | |
XM_017006637.1 | XP_016862126.1 | 1 - 2 | 430 | 69 | |
GenBank mRNA | HY178178.1 | - | 1 - 2 | 435 | 69 |