Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZKSCAN4
Entrez Gene ID
387032
Gene Name
zinc finger with KRAB and SCAN domains 4
Gene Aliases
P1P373C6, ZNF307, ZNF427, ZSCAN36
Chromosome Location
Chr.6: 28241697 - 28259312 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001304506.1 NP_001291435.1 2 - 3 219 91
NM_019110.4 NP_061983.2 2 - 3 884 91
XM_005249095.3 XP_005249152.1 4 - 5 928 91
XM_005249097.3 XP_005249154.1 4 - 5 624 91
XM_005249098.3 XP_005249155.1 2 - 3 542 91
XM_005249100.4 XP_005249157.2 2 - 3 275 91
XM_006715095.3 XP_006715158.1 3 - 4 401 91
XM_011514588.2 XP_011512890.1 2 - 3 784 91
XM_011514589.2 XP_011512891.1 2 - 3 340 91
XM_017010844.1 XP_016866333.1 4 - 5 1579 91
GenBank mRNA AK302017.1 - 2 - 3 200 91
AK315783.1 - 2 - 3 854 91
AY781778.1 - 2 - 3 854 91
BC014031.2 - 2 - 3 812 91
KU178905.1 - 2 - 3 568 91
KU178906.1 - 2 - 3 581 91

View Full Product Details