The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019593.3 | NP_062539.1 | 12 - 13 | 1366 | 135 |
XM_005260758.4 | XP_005260815.1 | 13 - 14 | 1517 | 135 | |
XM_005260761.1 | XP_005260818.1 | 11 - 12 | 1290 | 135 | |
XM_006723596.3 | XP_006723659.1 | 13 - 14 | 2290 | 135 | |
GenBank mRNA | AB037855.1 | - | 12 - 13 | 1357 | 135 |
AL833069.1 | - | 7 - 8 | 2562 | 135 | |
BC027588.2 | - | 12 - 13 | 1365 | 135 |