The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001297576.1 | NP_001284505.1 | 5 - 6 | 842 | 73 |
NM_001297577.1 | NP_001284506.1 | 4 - 5 | 640 | 73 | |
NM_001297578.1 | NP_001284507.1 | 3 - 4 | 469 | 73 | |
NM_003768.4 | NP_003759.1 | 3 - 4 | 535 | 73 | |
NR_123724.1 | 2 - 3 | 361 | 73 | ||
XM_006711599.2 | XP_006711662.1 | 3 - 4 | 439 | 73 | |
GenBank mRNA | AK095879.1 | - | 5 - 6 | 747 | 73 |
AK311762.1 | - | 3 - 4 | 464 | 73 | |
BC002426.2 | - | 3 - 4 | 411 | 73 | |
BC022554.1 | - | 3 - 4 | 522 | 73 | |
BT007252.1 | - | 2 - 3 | 329 | 73 | |
CR541733.1 | - | 2 - 3 | 329 | 73 | |
DA157938.1 | - | 3 - 4 | 405 | 73 | |
DA532423.1 | - | 3 - 4 | 519 | 73 | |
DA538845.1 | - | 2 - 3 | 299 | 73 | |
Y13736.1 | - | 3 - 4 | 398 | 73 |