Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RNASEH2B
Entrez Gene ID
79621
Gene Name
ribonuclease H2 subunit B
Gene Aliases
AGS2, DLEU8
Chromosome Location
Chr.13: 50909678 - 50970462 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024570.3 NP_078846.2 10 - 11 1216 95
XM_006719867.3 XP_006719930.1 10 - 11 905 95
XM_011535233.2 XP_011533535.1 11 - 12 1091 95
GenBank mRNA AK021774.1 - 10 - 11 1103 95
AK223340.1 - 10 - 11 1103 95
AY764036.1 - 10 - 11 1124 95
BC001397.2 - 10 - 11 926 95
BC005088.1 - 10 - 11 926 95
BC007332.2 - 10 - 11 926 95
BC010174.2 - 10 - 11 926 95
BC036744.1 - 10 - 11 1087 95

View Full Product Details