The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024109.3 | NP_077014.3 | 6 - 7 | 1082 | 77 |
NR_134608.1 | 5 - 6 | 1041 | 77 | ||
XM_005255570.3 | XP_005255627.1 | 6 - 7 | 1082 | 77 | |
XM_005255571.2 | XP_005255628.1 | 6 - 7 | 1082 | 77 | |
XM_011522657.2 | XP_011520959.1 | 6 - 7 | 1082 | 77 | |
XM_017023659.1 | XP_016879148.1 | 6 - 7 | 1082 | 77 | |
XM_017023660.1 | XP_016879149.1 | 7 - 8 | 1278 | 77 | |
XM_017023661.1 | XP_016879150.1 | 6 - 7 | 1082 | 77 | |
XM_017023662.1 | XP_016879151.1 | 5 - 6 | 996 | 77 | |
GenBank mRNA | AY203956.1 | - | 3 - 4 | 548 | 77 |
BC001908.1 | - | 5 - 6 | 990 | 77 |