The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319676.1 | NP_001306605.1 | 16 - 17 | 1912 | 116 |
NM_006615.2 | NP_006606.1 | 18 - 19 | 2101 | 116 | |
NM_016452.2 | NP_057536.1 | 17 - 18 | 2023 | 116 | |
XM_017000099.1 | XP_016855588.1 | 18 - 19 | 1831 | 116 | |
GenBank mRNA | AB038463.1 | - | 17 - 18 | 1992 | 116 |
AF022799.1 | - | 18 - 19 | 2101 | 116 | |
AK298024.1 | - | 16 - 17 | 1851 | 116 | |
BC027993.1 | - | 16 - 17 | 1863 | 116 |