The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319676.1 | NP_001306605.1 | 2 - 3 | 459 | 63 |
NM_006615.2 | NP_006606.1 | 4 - 5 | 648 | 63 | |
NM_016452.2 | NP_057536.1 | 4 - 5 | 648 | 63 | |
XM_011544017.1 | XP_011542319.1 | 4 - 5 | 632 | 63 | |
XM_011544018.1 | XP_011542320.1 | 4 - 5 | 632 | 63 | |
XM_011544019.2 | XP_011542321.1 | 2 - 3 | 554 | 63 | |
XM_011544020.1 | XP_011542322.1 | 4 - 5 | 378 | 63 | |
XM_017000098.1 | XP_016855587.1 | 4 - 5 | 632 | 63 | |
XM_017000099.1 | XP_016855588.1 | 4 - 5 | 378 | 63 | |
GenBank mRNA | AB038463.1 | - | 4 - 5 | 617 | 63 |
AF022799.1 | - | 4 - 5 | 648 | 63 | |
AK298024.1 | - | 2 - 3 | 398 | 63 | |
AK309039.1 | - | 4 - 5 | 378 | 63 | |
AK313629.1 | - | 4 - 5 | 63 | ||
BC027993.1 | - | 2 - 3 | 410 | 63 |