The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017424.2 | NP_001017424.1 | 2 - 3 | 727 | 86 |
NM_001017425.2 | NP_001017425.2 | 2 - 3 | 528 | 86 | |
NM_014217.3 | NP_055032.1 | 2 - 3 | 404 | 86 | |
XM_011509522.2 | XP_011507824.1 | 2 - 3 | 1930 | 86 | |
XM_011509523.1 | XP_011507825.1 | 2 - 3 | 332 | 86 | |
XM_011509524.2 | XP_011507826.1 | 2 - 3 | 379 | 86 | |
XM_017001248.1 | XP_016856737.1 | 3 - 4 | 1115 | 86 | |
XM_017001249.1 | XP_016856738.1 | 3 - 4 | 1066 | 86 | |
GenBank mRNA | AF004711.1 | - | 2 - 3 | 512 | 86 |
AF129399.1 | - | 1 - 2 | 314 | 86 | |
AF171068.1 | - | 2 - 3 | 328 | 86 | |
AK291483.1 | - | 2 - 3 | 508 | 86 | |
AK315249.1 | - | 3 - 4 | 494 | 86 | |
AU280191.1 | - | 2 - 3 | 470 | 86 | |
AY552980.2 | - | 2 - 3 | 414 | 86 | |
AY552981.1 | - | 2 - 3 | 419 | 86 | |
BC069462.1 | - | 2 - 3 | 328 | 86 | |
BC101693.1 | - | 2 - 3 | 373 | 86 | |
BC101695.1 | - | 2 - 3 | 373 | 86 | |
BC143586.1 | - | 2 - 3 | 374 | 86 | |
CD625301.1 | - | 2 - 3 | 343 | 86 | |
EF165334.1 | - | 2 - 3 | 461 | 86 | |
EF165335.1 | - | 2 - 3 | 426 | 86 | |
KF182338.1 | - | 2 - 3 | 363 | 86 | |
KF182339.1 | - | 2 - 3 | 337 | 86 |