The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017424.2 | NP_001017424.1 | 4 - 5 | 1002 | 114 |
NM_001017425.2 | NP_001017425.2 | 4 - 5 | 803 | 114 | |
NM_014217.3 | NP_055032.1 | 4 - 5 | 679 | 114 | |
XM_011509522.2 | XP_011507824.1 | 4 - 5 | 2205 | 114 | |
XM_011509523.1 | XP_011507825.1 | 4 - 5 | 607 | 114 | |
XM_011509524.2 | XP_011507826.1 | 4 - 5 | 654 | 114 | |
XM_017001248.1 | XP_016856737.1 | 5 - 6 | 1390 | 114 | |
XM_017001249.1 | XP_016856738.1 | 5 - 6 | 1341 | 114 | |
GenBank mRNA | AF004711.1 | - | 4 - 5 | 787 | 114 |
AF129399.1 | - | 3 - 4 | 589 | 114 | |
AF171068.1 | - | 4 - 5 | 603 | 114 | |
AK123409.1 | - | 3 - 4 | 573 | 114 | |
AK291483.1 | - | 4 - 5 | 783 | 114 | |
AK294780.1 | - | 3 - 4 | 667 | 114 | |
AK315249.1 | - | 5 - 6 | 769 | 114 | |
AY552980.2 | - | 4 - 5 | 689 | 114 | |
BC069462.1 | - | 4 - 5 | 603 | 114 | |
BC101693.1 | - | 4 - 5 | 648 | 114 | |
BC101695.1 | - | 4 - 5 | 648 | 114 | |
BC143586.1 | - | 4 - 5 | 649 | 114 | |
EF165334.1 | - | 4 - 5 | 736 | 114 | |
EF165335.1 | - | 4 - 5 | 701 | 114 | |
KF182338.1 | - | 4 - 5 | 638 | 114 | |
KF182339.1 | - | 4 - 5 | 612 | 114 |