The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017780.3 | NP_060250.2 | 32 - 33 | 7424 | 82 |
XM_011517553.2 | XP_011515855.1 | 33 - 34 | 7535 | 82 | |
XM_011517554.2 | XP_011515856.1 | 33 - 34 | 7513 | 82 | |
XM_011517555.2 | XP_011515857.1 | 33 - 34 | 7532 | 82 | |
XM_017013612.1 | XP_016869101.1 | 33 - 34 | 7589 | 82 | |
XM_017013613.1 | XP_016869102.1 | 32 - 33 | 7442 | 82 | |
GenBank mRNA | AB037837.1 | - | 28 - 29 | 4646 | 82 |
AK295751.1 | - | 11 - 12 | 2019 | 82 |