Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHD7
Entrez Gene ID
55636
Gene Name
chromodomain helicase DNA binding protein 7
Gene Aliases
CRG, HH5, IS3, KAL5
Chromosome Location
Chr.8: 60678744 - 60868028 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017780.3 NP_060250.2 35 - 36 8314 94
XM_011517553.2 XP_011515855.1 36 - 37 8425 94
XM_011517554.2 XP_011515856.1 36 - 37 8403 94
XM_011517555.2 XP_011515857.1 36 - 37 8422 94
XM_017013612.1 XP_016869101.1 36 - 37 8479 94
XM_017013613.1 XP_016869102.1 35 - 36 8332 94
GenBank mRNA AB037837.1 - 31 - 32 5536 94
AK000364.1 - 4 - 5 912 94
AK295751.1 - 14 - 15 2909 94
BC051264.1 - 1 - 2 39 94

View Full Product Details