The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243142.1 | NP_001230071.1 | 10 - 11 | 1490 | 152 |
NM_001243143.1 | NP_001230072.1 | 10 - 11 | 1451 | 152 | |
NM_001301136.1 | NP_001288065.1 | 10 - 11 | 1493 | 152 | |
NM_016359.4 | NP_057443.2 | 10 - 11 | 1496 | 152 | |
NM_018454.7 | NP_060924.4 | 10 - 11 | 1493 | 152 | |
XM_005254428.2 | XP_005254485.1 | 10 - 11 | 1365 | 152 | |
XM_005254430.4 | XP_005254487.1 | 10 - 11 | 1287 | 152 | |
XM_006720559.2 | XP_006720622.1 | 10 - 11 | 1368 | 152 | |
XM_006720560.3 | XP_006720623.1 | 10 - 11 | 1464 | 152 | |
XM_006720561.2 | XP_006720624.1 | 10 - 11 | 1282 | 152 | |
XM_017022295.1 | XP_016877784.1 | 10 - 11 | 2160 | 152 | |
GenBank mRNA | AF090915.1 | - | 10 - 11 | 1299 | 152 |
AF290612.1 | - | 10 - 11 | 1300 | 152 | |
AF305711.1 | - | 10 - 11 | 1229 | 152 | |
AK023483.1 | - | 10 - 11 | 1303 | 152 | |
AK222819.1 | - | 10 - 11 | 1317 | 152 | |
AK225703.1 | - | 10 - 11 | 1307 | 152 | |
AK290879.1 | - | 10 - 11 | 1320 | 152 | |
AK307728.1 | - | 10 - 11 | 1290 | 152 | |
AL833611.1 | - | 10 - 11 | 1263 | 152 | |
BC001308.1 | - | 10 - 11 | 1296 | 152 | |
BC010838.1 | - | 8 - 9 | 1037 | 152 | |
BC012887.1 | - | 10 - 11 | 1274 | 152 | |
BC024772.1 | - | 10 - 11 | 1298 | 152 |