Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TROAP
Entrez Gene ID
10024
Gene Name
trophinin associated protein
Gene Aliases
TASTIN
Chromosome Location
Chr.12: 49322135 - 49331733 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005480.3 NP_005471.3 13 - 14 2267 83
GenBank mRNA BC032583.1 - 13 - 14 2179 83
U04810.2 - 13 - 14 2210 83

View Full Product Details