Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TROAP
Entrez Gene ID
10024
Gene Name
trophinin associated protein
Gene Aliases
TASTIN
Chromosome Location
Chr.12: 49322135 - 49331733 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005480.3 NP_005471.3 3 - 4 503 108
XM_006719181.1 XP_006719244.1 3 - 4 447 108
XM_011537723.1 XP_011536025.1 3 - 4 447 108
GenBank mRNA AK299302.1 - 3 - 4 408 108
AK315211.1 - 3 - 4 447 108
BC032583.1 - 3 - 4 415 108
U04810.2 - 3 - 4 446 108

View Full Product Details