The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005480.3 | NP_005471.3 | 6 - 7 | 885 | 77 |
XM_006719181.1 | XP_006719244.1 | 6 - 7 | 829 | 77 | |
XM_011537723.1 | XP_011536025.1 | 6 - 7 | 829 | 77 | |
XM_011537724.1 | XP_011536026.1 | 5 - 6 | 625 | 77 | |
GenBank mRNA | AK296811.1 | - | 6 - 7 | 772 | 77 |
AK299302.1 | - | 6 - 7 | 692 | 77 | |
AK310709.1 | - | 5 - 6 | 623 | 77 | |
AK315211.1 | - | 6 - 7 | 829 | 77 | |
BC032583.1 | - | 6 - 7 | 797 | 77 | |
U04810.2 | - | 6 - 7 | 828 | 77 |