Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TROAP
Entrez Gene ID
10024
Gene Name
trophinin associated protein
Gene Aliases
TASTIN
Chromosome Location
Chr.12: 49322135 - 49331733 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005480.3 NP_005471.3 7 - 8 935 71
XM_006719181.1 XP_006719244.1 7 - 8 879 71
XM_011537723.1 XP_011536025.1 7 - 8 879 71
XM_011537724.1 XP_011536026.1 6 - 7 675 71
GenBank mRNA AK296811.1 - 7 - 8 822 71
AK299302.1 - 7 - 8 742 71
AK309716.1 - 4 - 5 356 71
AK310709.1 - 6 - 7 673 71
AK315211.1 - 7 - 8 879 71
BC032583.1 - 7 - 8 847 71
U04810.2 - 7 - 8 878 71

View Full Product Details