The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006772.2 | NP_006763.2 | 7 - 8 | 955 | 65 |
GenBank mRNA | AK094225.1 | - | 7 - 8 | 955 | 65 |
AK307888.1 | - | 7 - 8 | 770 | 65 |