The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282535.1 | NP_001269464.1 | 2 - 3 | 1641 | 69 |
GenBank mRNA | AL136878.1 | - | 2 - 3 | 1533 | 69 |