Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ITGA10
Entrez Gene ID
8515
Gene Name
integrin subunit alpha 10
Gene Aliases
PRO827
Chromosome Location
Chr.1: 145891208 - 145910189 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001303040.1 NP_001289969.1 19 - 20 2607 70
NM_001303041.1 NP_001289970.1 18 - 19 2495 70
NM_003637.4 NP_003628.2 22 - 23 2924 70
XM_005277436.3 XP_005277493.1 21 - 22 2675 70
XM_011510083.2 XP_011508385.1 18 - 19 2355 70
XM_011510084.1 XP_011508386.1 11 - 12 1352 70
XM_017002622.1 XP_016858111.1 20 - 21 2881 70
XM_017002623.1 XP_016858112.1 20 - 21 2878 70
XM_017002624.1 XP_016858113.1 20 - 21 2881 70
XM_017002625.1 XP_016858114.1 20 - 21 2881 70
XM_017002626.1 XP_016858115.1 18 - 19 2638 70
XM_017002628.1 XP_016858117.1 16 - 17 2331 70
GenBank mRNA AF112345.1 - 22 - 23 2825 70
AK304155.1 - 19 - 20 2463 70
AK304183.1 - 21 - 22 2754 70
AK314255.1 - 22 - 23 2773 70
BC140831.1 - 18 - 19 2370 70
BC144637.1 - 18 - 19 2370 70

View Full Product Details