Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ITGA10
Entrez Gene ID
8515
Gene Name
integrin subunit alpha 10
Gene Aliases
PRO827
Chromosome Location
Chr.1: 145891208 - 145910189 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003637.4 NP_003628.2 4 - 5 544 102
XM_005277436.3 XP_005277493.1 3 - 4 295 102
XM_017002622.1 XP_016858111.1 2 - 3 501 102
XM_017002623.1 XP_016858112.1 2 - 3 501 102
XM_017002624.1 XP_016858113.1 2 - 3 501 102
XM_017002625.1 XP_016858114.1 2 - 3 501 102
XM_017002627.1 XP_016858116.1 2 - 3 501 102
GenBank mRNA AF074015.1 - 4 - 5 390 102
AF112345.1 - 4 - 5 445 102
AK304183.1 - 3 - 4 374 102
AK314255.1 - 4 - 5 393 102

View Full Product Details