The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001079843.2 | NP_001073312.1 | 14 - 15 | 3379 | 57 |
NM_017766.4 | NP_060236.3 | 14 - 15 | 3379 | 57 | |
XM_005263479.3 | XP_005263536.1 | 12 - 13 | 3243 | 57 | |
XM_011541635.2 | XP_011539937.1 | 12 - 13 | 3242 | 57 | |
XM_017001539.1 | XP_016857028.1 | 12 - 13 | 3243 | 57 | |
XM_017001540.1 | XP_016857029.1 | 14 - 15 | 3353 | 57 | |
XM_017001541.1 | XP_016857030.1 | 14 - 15 | 3415 | 57 | |
GenBank mRNA | AK000328.1 | - | 10 - 11 | 2693 | 57 |
AK092289.1 | - | 14 - 15 | 3353 | 57 | |
AK225324.1 | - | 10 - 11 | 2693 | 57 | |
BC004410.1 | - | 5 - 6 | 1378 | 57 | |
BC051883.2 | - | 11 - 12 | 3049 | 57 | |
DQ217660.1 | - | 14 - 15 | 3351 | 57 |