The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001079843.2 | NP_001073312.1 | 8 - 9 | 1846 | 58 |
NM_017766.4 | NP_060236.3 | 8 - 9 | 1846 | 58 | |
XM_005263479.3 | XP_005263536.1 | 6 - 7 | 1710 | 58 | |
XM_011541635.2 | XP_011539937.1 | 6 - 7 | 1709 | 58 | |
XM_017001539.1 | XP_016857028.1 | 6 - 7 | 1710 | 58 | |
XM_017001540.1 | XP_016857029.1 | 8 - 9 | 1820 | 58 | |
XM_017001541.1 | XP_016857030.1 | 8 - 9 | 1882 | 58 | |
GenBank mRNA | AK000328.1 | - | 4 - 5 | 1160 | 58 |
AK225324.1 | - | 4 - 5 | 1160 | 58 | |
AK293839.1 | - | 6 - 7 | 1678 | 58 | |
BC051883.2 | - | 5 - 6 | 1516 | 58 | |
DQ217660.1 | - | 8 - 9 | 1818 | 58 |