Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ATG16L1
- Entrez Gene ID
-
55054
- Gene Name
- autophagy related 16 like 1
- Gene Aliases
-
APG16L, ATG16A, ATG16L, IBD10, WDR30
- Chromosome Location
-
Chr.2: 233251571 - 233295674 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK000897.1 |
- |
4 - 5 |
531 |
64 |
View Full Product Details