The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004984.2 | NP_004975.2 | 14 - 15 | 1782 | 67 |
GenBank mRNA | AB210045.1 | - | 14 - 15 | 1607 | 67 |
AB290174.1 | - | 14 - 15 | 1848 | 67 | |
AK294881.1 | - | 11 - 12 | 1535 | 67 | |
AK315202.1 | - | 14 - 15 | 1802 | 67 | |
BC146670.1 | - | 14 - 15 | 1785 | 67 | |
BC150208.1 | - | 14 - 15 | 1785 | 67 | |
U06698.1 | - | 14 - 15 | 1722 | 67 |