Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLCG1
Entrez Gene ID
5335
Gene Name
phospholipase C gamma 1
Gene Aliases
NCKAP3, PLC-II, PLC1, PLC148, PLCgamma1
Chromosome Location
Chr.20: 41137519 - 41175721 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002660.2 NP_002651.2 19 - 20 2419 56
NM_182811.1 NP_877963.1 19 - 20 2419 56
XM_005260438.2 XP_005260495.1 19 - 20 2421 56
XM_011528865.1 XP_011527167.1 19 - 20 2286 56
XM_011528867.2 XP_011527169.1 19 - 20 2419 56
GenBank mRNA AB210028.1 - 19 - 20 2703 56
AK297370.1 - 8 - 9 1169 56
AL110247.1 - 1 - 2 153 56
BC136466.1 - 19 - 20 2386 56
BC144136.1 - 19 - 20 2386 56
M34667.1 - 19 - 20 2374 56

View Full Product Details