The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002660.2 | NP_002651.2 | 20 - 21 | 2497 | 77 |
NM_182811.1 | NP_877963.1 | 20 - 21 | 2497 | 77 | |
XM_005260438.2 | XP_005260495.1 | 20 - 21 | 2499 | 77 | |
XM_011528865.1 | XP_011527167.1 | 20 - 21 | 2364 | 77 | |
XM_011528867.2 | XP_011527169.1 | 20 - 21 | 2497 | 77 | |
GenBank mRNA | AB210028.1 | - | 20 - 21 | 2781 | 77 |
AL110247.1 | - | 2 - 3 | 231 | 77 | |
BC136466.1 | - | 20 - 21 | 2464 | 77 | |
BC144136.1 | - | 20 - 21 | 2464 | 77 | |
M34667.1 | - | 20 - 21 | 2452 | 77 |